Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
Chromosome Location
Chr.11: 61299447 - 61333212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001923.4 NP_001914.3 7 - 8 1160 66
GenBank mRNA AJ002955.1 - 7 - 8 1025 66
AK294493.1 - 9 - 10 1289 66
AK299650.1 - 6 - 7 609 66
AK302924.1 - 5 - 6 558 66
AK311476.1 - 5 - 6 676 66
AK312436.1 - 7 - 8 982 66
AL831958.1 - 7 - 8 1142 66
BC011686.2 - 7 - 8 989 66
BC021044.1 - 7 - 8 977 66
BC050530.1 - 7 - 8 1053 66
BC051764.2 - 7 - 8 1043 66
L40326.1 - 7 - 8 954 66
U18299.1 - 7 - 8 1016 66
U32986.1 - 7 - 8 1025 66

View Full Product Details