The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021248.2 | NP_067071.1 | 5 - 6 | 1480 | 71 |
XM_011528994.2 | XP_011527296.1 | 5 - 6 | 1439 | 71 | |
XM_011528995.1 | XP_011527297.1 | 5 - 6 | 1244 | 71 | |
GenBank mRNA | AK289584.1 | - | 5 - 6 | 1480 | 71 |
BC031232.1 | - | 3 - 4 | 705 | 71 | |
BC136526.1 | - | 4 - 5 | 1229 | 71 |