The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015478.6 | NP_056293.4 | 6 - 7 | 893 | 61 |
NM_032107.4 | NP_115479.4 | 9 - 10 | 1064 | 61 | |
GenBank mRNA | AB014581.1 | - | 2 - 3 | 143 | 61 |
AK299199.1 | - | 9 - 10 | 1034 | 61 | |
AK310345.1 | - | 6 - 7 | 893 | 61 | |
BC039820.1 | - | 6 - 7 | 808 | 61 |