Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
Chromosome Location
Chr.20: 63235883 - 63240507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139317.2 NP_647478.1 5 - 6 861 114
GenBank mRNA AF301009.1 - 5 - 6 647 114
AY358836.1 - 5 - 6 861 114
BC014475.1 - 5 - 6 795 114

View Full Product Details