The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025096.1 | NP_001020267.1 | 4 - 5 | 588 | 139 |
NM_001025097.1 | NP_001020268.1 | 4 - 5 | 474 | 139 | |
NM_001308188.1 | NP_001295117.1 | 5 - 6 | 658 | 139 | |
NM_144662.2 | NP_653263.2 | 4 - 5 | 606 | 139 | |
XM_011525818.1 | XP_011524120.1 | 5 - 6 | 541 | 139 | |
GenBank mRNA | BC021713.1 | - | 5 - 6 | 598 | 139 |
BC025389.1 | - | 4 - 5 | 584 | 139 | |
BC025393.1 | - | 4 - 5 | 462 | 139 | |
BC028371.1 | - | 4 - 5 | 462 | 139 | |
BC028686.1 | - | 4 - 5 | 462 | 139 | |
BC042820.1 | - | 4 - 5 | 588 | 139 | |
BC047355.1 | - | 4 - 5 | 580 | 139 |