Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
Chromosome Location
Chr.3: 148697871 - 148743003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009585.3 NP_033611.1 1 - 2 256 148
GenBank mRNA BC022447.1 - 1 - 2 224 148

View Full Product Details