The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144718.3 | NP_653319.1 | 15 - 16 | 2611 | 93 |
XM_011512461.2 | XP_011510763.1 | 15 - 16 | 2368 | 93 | |
XM_017005775.1 | XP_016861264.1 | 15 - 16 | 2368 | 93 | |
XM_017005776.1 | XP_016861265.1 | 15 - 16 | 2505 | 93 | |
XM_017005777.1 | XP_016861266.1 | 15 - 16 | 2428 | 93 | |
XM_017005778.1 | XP_016861267.1 | 15 - 16 | 2612 | 93 | |
GenBank mRNA | AK129575.1 | - | 5 - 6 | 1140 | 93 |
AL832148.1 | - | 15 - 16 | 2580 | 93 | |
AL832327.2 | - | 15 - 16 | 4276 | 93 | |
AY099107.1 | - | 15 - 16 | 2531 | 93 | |
BC019232.1 | - | 1 - 2 | 107 | 93 | |
BC036951.1 | - | 15 - 16 | 2552 | 93 | |
BC038595.1 | - | 3 - 4 | 734 | 93 |