The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144718.3 | NP_653319.1 | 8 - 9 | 1043 | 68 |
XM_011512461.2 | XP_011510763.1 | 8 - 9 | 800 | 68 | |
XM_017005775.1 | XP_016861264.1 | 8 - 9 | 800 | 68 | |
XM_017005776.1 | XP_016861265.1 | 8 - 9 | 937 | 68 | |
XM_017005777.1 | XP_016861266.1 | 8 - 9 | 860 | 68 | |
XM_017005778.1 | XP_016861267.1 | 8 - 9 | 1044 | 68 | |
GenBank mRNA | AK126897.1 | - | 4 - 5 | 449 | 68 |
AL832148.1 | - | 8 - 9 | 1012 | 68 | |
AL832327.2 | - | 8 - 9 | 826 | 68 | |
AY099107.1 | - | 8 - 9 | 963 | 68 | |
BC036951.1 | - | 8 - 9 | 984 | 68 |