The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001690.3 | NP_001681.2 | 2 - 3 | 191 | 101 |
GenBank mRNA | AF113129.1 | - | 2 - 3 | 118 | 101 |
AK314779.1 | - | 2 - 3 | 154 | 101 | |
AL514316.3 | - | 2 - 3 | 161 | 101 | |
BC013138.1 | - | 2 - 3 | 139 | 101 | |
BT006672.1 | - | 1 - 2 | 83 | 101 | |
DC350361.1 | - | 2 - 3 | 191 | 101 | |
L09235.1 | - | 1 - 2 | 98 | 101 |