The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282959.1 | NP_001269888.1 | 21 - 22 | 3151 | 81 |
NM_001282960.1 | NP_001269889.1 | 21 - 22 | 3145 | 81 | |
NM_018237.3 | NP_060707.2 | 22 - 23 | 3190 | 81 | |
NR_104262.1 | 21 - 22 | 2985 | 81 | ||
GenBank mRNA | AF465616.1 | - | 22 - 23 | 3120 | 81 |
AK299307.1 | - | 21 - 22 | 3050 | 81 | |
AY249140.1 | - | 22 - 23 | 3098 | 81 | |
BC130626.1 | - | 22 - 23 | 3079 | 81 | |
BC132725.1 | - | 22 - 23 | 3079 | 81 | |
BC144118.1 | - | 21 - 22 | 2874 | 81 | |
BM905703.1 | - | - | 567 | 81 | |
CB136079.1 | - | 2 - 3 | 164 | 81 |