The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198696.1 | NP_001185625.1 | 5 - 6 | 726 | 76 |
NM_032797.5 | NP_116186.1 | 5 - 6 | 723 | 76 | |
GenBank mRNA | AF337957.1 | - | 4 - 5 | 527 | 76 |
AF506757.1 | - | 5 - 6 | 723 | 76 | |
AK027403.1 | - | 5 - 6 | 602 | 76 | |
AK127353.1 | - | 5 - 6 | 2478 | 76 | |
BC006121.1 | - | 4 - 5 | 531 | 76 | |
BC023601.2 | - | 5 - 6 | 710 | 76 | |
BX537621.1 | - | 3 - 4 | 323 | 76 |