The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017977.2 | NP_001017977.1 | 10 - 11 | 1734 | 75 |
XM_005245333.4 | XP_005245390.1 | 10 - 11 | 1675 | 75 | |
XM_017001781.1 | XP_016857270.1 | 10 - 11 | 1533 | 75 | |
GenBank mRNA | AK226181.1 | - | 10 - 11 | 1585 | 75 |
AY766164.1 | - | 10 - 11 | 1474 | 75 | |
BC025262.1 | - | 2 - 3 | 397 | 75 | |
DQ768089.1 | - | 10 - 11 | 1398 | 75 |