Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
Chromosome Location
Chr.1: 167936247 - 168075843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017977.2 NP_001017977.1 2 - 3 510 148
NM_001198956.1 NP_001185885.1 2 - 3 510 148
NM_018442.3 NP_060912.2 2 - 3 510 148
XM_005245331.4 XP_005245388.1 2 - 3 451 148
XM_005245332.4 XP_005245389.1 2 - 3 451 148
XM_005245333.4 XP_005245390.1 2 - 3 451 148
XM_017001779.1 XP_016857268.1 2 - 3 323 148
XM_017001780.1 XP_016857269.1 2 - 3 259 148
XM_017001781.1 XP_016857270.1 2 - 3 309 148
GenBank mRNA AF116725.1 - 2 - 3 251 148
AK093970.1 - 2 - 3 396 148
AK226181.1 - 2 - 3 361 148
AY766164.1 - 2 - 3 250 148
DQ768089.1 - 2 - 3 174 148

View Full Product Details