The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017977.2 | NP_001017977.1 | 5 - 6 | 898 | 68 |
NM_001198956.1 | NP_001185885.1 | 5 - 6 | 898 | 68 | |
NM_001198957.1 | NP_001185886.1 | 4 - 5 | 805 | 68 | |
NM_018442.3 | NP_060912.2 | 5 - 6 | 898 | 68 | |
XM_005245331.4 | XP_005245388.1 | 5 - 6 | 839 | 68 | |
XM_005245332.4 | XP_005245389.1 | 5 - 6 | 839 | 68 | |
XM_005245333.4 | XP_005245390.1 | 5 - 6 | 839 | 68 | |
XM_017001779.1 | XP_016857268.1 | 5 - 6 | 711 | 68 | |
XM_017001780.1 | XP_016857269.1 | 5 - 6 | 647 | 68 | |
XM_017001781.1 | XP_016857270.1 | 5 - 6 | 697 | 68 | |
GenBank mRNA | AF116725.1 | - | 5 - 6 | 639 | 68 |
AF124434.1 | - | 3 - 4 | 334 | 68 | |
AF150734.1 | - | 2 - 3 | 189 | 68 | |
AK093970.1 | - | 5 - 6 | 784 | 68 | |
AK226181.1 | - | 5 - 6 | 749 | 68 | |
AK297847.1 | - | 4 - 5 | 665 | 68 | |
AY766164.1 | - | 5 - 6 | 638 | 68 | |
DQ768089.1 | - | 5 - 6 | 562 | 68 |