The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130704.1 | NP_001124176.1 | 2 - 3 | 305 | 77 |
NM_001278125.1 | NP_001265054.1 | 4 - 5 | 472 | 77 | |
NM_001278126.1 | NP_001265055.1 | 3 - 4 | 411 | 77 | |
NM_014269.4 | NP_055084.3 | 3 - 4 | 411 | 77 | |
XM_011531556.1 | XP_011529858.1 | 3 - 4 | 397 | 77 | |
XM_011531557.1 | XP_011529859.1 | 3 - 4 | 397 | 77 | |
XM_011531559.1 | XP_011529861.1 | 3 - 4 | 394 | 77 | |
GenBank mRNA | AK292410.1 | - | 2 - 3 | 260 | 77 |
DB078034.1 | - | 3 - 4 | 366 | 77 | |
DB078125.1 | - | 3 - 4 | 365 | 77 | |
DB078627.1 | - | 4 - 5 | 427 | 77 | |
DB080702.1 | - | 2 - 3 | 259 | 77 |