The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130704.1 | NP_001124176.1 | 3 - 4 | 388 | 95 |
NM_001278125.1 | NP_001265054.1 | 5 - 6 | 555 | 95 | |
NM_014269.4 | NP_055084.3 | 4 - 5 | 494 | 95 | |
GenBank mRNA | AK292410.1 | - | 3 - 4 | 343 | 95 |
DB078034.1 | - | 4 - 5 | 449 | 95 | |
DB078627.1 | - | 5 - 6 | 510 | 95 | |
DB080702.1 | - | 3 - 4 | 342 | 95 |