Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 11 - 12 2010 103
XM_005244966.4 XP_005245023.2 14 - 15 2099 103
XM_005244967.4 XP_005245024.2 11 - 12 1936 103
XM_005244968.4 XP_005245025.2 11 - 12 1876 103
XM_006711212.3 XP_006711275.1 14 - 15 2385 103
XM_006711213.3 XP_006711276.2 13 - 14 2105 103
XM_006711214.3 XP_006711277.2 14 - 15 2159 103
XM_006711215.3 XP_006711278.2 12 - 13 2024 103
XM_006711217.3 XP_006711280.2 14 - 15 2159 103
XM_006711219.3 XP_006711282.2 11 - 12 1937 103
XM_006711221.2 XP_006711284.1 14 - 15 2483 103
XM_006711222.3 XP_006711285.1 14 - 15 2468 103
XM_011509295.2 XP_011507597.1 14 - 15 2159 103
XM_011509296.2 XP_011507598.1 13 - 14 2078 103
XM_011509297.2 XP_011507599.1 16 - 17 2985 103
XM_011509298.2 XP_011507600.1 14 - 15 2268 103
XM_011509299.2 XP_011507601.2 14 - 15 2274 103
XM_011509301.2 XP_011507603.1 14 - 15 6164 103
XM_011509304.2 XP_011507606.1 14 - 15 2113 103
XM_017000687.1 XP_016856176.1 14 - 15 2159 103
XM_017000688.1 XP_016856177.1 13 - 14 2105 103
XM_017000689.1 XP_016856178.1 12 - 13 1964 103
XM_017000690.1 XP_016856179.1 14 - 15 6104 103
XM_017000691.1 XP_016856180.1 9 - 10 4557 103
XM_017000692.1 XP_016856181.1 9 - 10 4497 103
GenBank mRNA AB023186.1 - 11 - 12 1997 103
BC152475.1 - 11 - 12 1997 103

View Full Product Details