Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 13 - 14 2245 62
XM_005244966.4 XP_005245023.2 16 - 17 2334 62
XM_005244967.4 XP_005245024.2 13 - 14 2171 62
XM_005244968.4 XP_005245025.2 13 - 14 2111 62
XM_006711212.3 XP_006711275.1 16 - 17 2620 62
XM_006711213.3 XP_006711276.2 15 - 16 2340 62
XM_006711214.3 XP_006711277.2 16 - 17 2394 62
XM_006711215.3 XP_006711278.2 14 - 15 2259 62
XM_006711217.3 XP_006711280.2 16 - 17 2394 62
XM_006711219.3 XP_006711282.2 13 - 14 2172 62
XM_006711221.2 XP_006711284.1 16 - 17 2718 62
XM_006711222.3 XP_006711285.1 16 - 17 2703 62
XM_011509295.2 XP_011507597.1 16 - 17 2394 62
XM_011509296.2 XP_011507598.1 15 - 16 2313 62
XM_011509297.2 XP_011507599.1 18 - 19 3220 62
XM_011509298.2 XP_011507600.1 16 - 17 2503 62
XM_011509299.2 XP_011507601.2 16 - 17 2509 62
XM_011509301.2 XP_011507603.1 16 - 17 6399 62
XM_011509304.2 XP_011507606.1 16 - 17 2348 62
XM_017000687.1 XP_016856176.1 16 - 17 2394 62
XM_017000688.1 XP_016856177.1 15 - 16 2340 62
XM_017000689.1 XP_016856178.1 14 - 15 2199 62
XM_017000690.1 XP_016856179.1 16 - 17 6339 62
XM_017000691.1 XP_016856180.1 11 - 12 4792 62
XM_017000692.1 XP_016856181.1 11 - 12 4732 62
GenBank mRNA AB023186.1 - 13 - 14 2232 62
BC152475.1 - 13 - 14 2232 62

View Full Product Details