Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 22 - 23 3486 76
XM_005244966.4 XP_005245023.2 26 - 27 3728 76
XM_005244967.4 XP_005245024.2 22 - 23 3415 76
XM_005244968.4 XP_005245025.2 22 - 23 3352 76
XM_006711212.3 XP_006711275.1 26 - 27 4014 76
XM_006711213.3 XP_006711276.2 25 - 26 3734 76
XM_006711215.3 XP_006711278.2 24 - 25 3653 76
XM_006711217.3 XP_006711280.2 25 - 26 3638 76
XM_006711219.3 XP_006711282.2 23 - 24 3566 76
XM_006711221.2 XP_006711284.1 26 - 27 4112 76
XM_006711222.3 XP_006711285.1 26 - 27 4097 76
XM_011509295.2 XP_011507597.1 26 - 27 3785 76
XM_011509296.2 XP_011507598.1 25 - 26 3707 76
XM_011509297.2 XP_011507599.1 28 - 29 4614 76
XM_011509298.2 XP_011507600.1 26 - 27 3897 76
XM_011509299.2 XP_011507601.2 26 - 27 3903 76
XM_011509301.2 XP_011507603.1 26 - 27 7793 76
XM_011509304.2 XP_011507606.1 26 - 27 3742 76
XM_017000689.1 XP_016856178.1 24 - 25 3593 76
XM_017000690.1 XP_016856179.1 26 - 27 7733 76
XM_017000691.1 XP_016856180.1 21 - 22 6186 76
XM_017000692.1 XP_016856181.1 21 - 22 6126 76
GenBank mRNA AB023186.1 - 22 - 23 3473 76
BC152475.1 - 22 - 23 3473 76

View Full Product Details