Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 2 - 3 317 89
XM_005244966.4 XP_005245023.2 2 - 3 184 89
XM_005244967.4 XP_005245024.2 2 - 3 183 89
XM_005244968.4 XP_005245025.2 2 - 3 183 89
XM_006711212.3 XP_006711275.1 2 - 3 410 89
XM_006711213.3 XP_006711276.2 2 - 3 184 89
XM_006711214.3 XP_006711277.2 2 - 3 184 89
XM_006711215.3 XP_006711278.2 2 - 3 184 89
XM_006711217.3 XP_006711280.2 2 - 3 184 89
XM_006711219.3 XP_006711282.2 2 - 3 184 89
XM_006711221.2 XP_006711284.1 2 - 3 508 89
XM_006711222.3 XP_006711285.1 2 - 3 493 89
XM_011509295.2 XP_011507597.1 2 - 3 184 89
XM_011509296.2 XP_011507598.1 2 - 3 184 89
XM_011509297.2 XP_011507599.1 4 - 5 1010 89
XM_011509298.2 XP_011507600.1 2 - 3 293 89
XM_011509299.2 XP_011507601.2 2 - 3 299 89
XM_011509301.2 XP_011507603.1 2 - 3 4189 89
XM_011509304.2 XP_011507606.1 2 - 3 138 89
XM_017000687.1 XP_016856176.1 2 - 3 184 89
XM_017000688.1 XP_016856177.1 2 - 3 184 89
XM_017000689.1 XP_016856178.1 2 - 3 184 89
XM_017000690.1 XP_016856179.1 2 - 3 4189 89
GenBank mRNA AB023186.1 - 2 - 3 304 89
BC152475.1 - 2 - 3 304 89
BX954305.1 - 2 - 3 318 89

View Full Product Details