Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
Chromosome Location
Chr.4: 170060222 - 170091699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286682.1 NP_001273611.1 1 - 2 75 67
NM_182662.1 NP_872603.1 1 - 2 75 67
GenBank mRNA AK055952.1 - 1 - 2 75 67
BC031068.1 - 1 - 2 68 67

View Full Product Details