Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPY5R
Entrez Gene ID
4889
Gene Name
neuropeptide Y receptor Y5
Gene Aliases
NPY5-R, NPYR5, NPYY5-R
Chromosome Location
Chr.4: 163336968 - 163352277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006174.3 NP_006165.1 2 - 3 193 79
XM_005263038.3 XP_005263095.1 2 - 3 7072 79
XM_011532015.2 XP_011530317.1 2 - 3 7072 79
GenBank mRNA BC042416.1 - 2 - 3 101 79

View Full Product Details