Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
Chromosome Location
Chr.1: 102876467 - 103108580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190709.1 NP_001177638.1 20 - 21 2204 98
NM_001854.3 NP_001845.3 21 - 22 2321 98
NM_080629.2 NP_542196.2 21 - 22 2357 98
NM_080630.3 NP_542197.3 19 - 20 1973 98
NR_134980.1 21 - 22 2321 98
XM_017000334.1 XP_016855823.1 22 - 23 2557 98
XM_017000335.1 XP_016855824.1 22 - 23 2551 98
XM_017000336.1 XP_016855825.1 22 - 23 2558 98
XM_017000337.1 XP_016855826.1 15 - 16 1035 98
GenBank mRNA AB208844.1 - 17 - 18 1321 98
BC117697.1 - 19 - 20 1956 98
J04177.1 - 21 - 22 2164 98

View Full Product Details