The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190709.1 | NP_001177638.1 | 64 - 65 | 5238 | 112 |
NM_001854.3 | NP_001845.3 | 65 - 66 | 5355 | 112 | |
NM_080629.2 | NP_542196.2 | 65 - 66 | 5391 | 112 | |
NM_080630.3 | NP_542197.3 | 63 - 64 | 5007 | 112 | |
NR_134980.1 | 66 - 67 | 5371 | 112 | ||
XM_017000334.1 | XP_016855823.1 | 66 - 67 | 5591 | 112 | |
XM_017000335.1 | XP_016855824.1 | 66 - 67 | 5585 | 112 | |
XM_017000337.1 | XP_016855826.1 | 59 - 60 | 4069 | 112 | |
GenBank mRNA | AB208844.1 | - | 62 - 63 | 4371 | 112 |
AK299025.1 | - | 3 - 4 | 288 | 112 | |
BC117697.1 | - | 63 - 64 | 4990 | 112 | |
BU159588.1 | - | 6 - 7 | 582 | 112 | |
J04177.1 | - | 65 - 66 | 5198 | 112 |