The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288952.1 | NP_001275881.1 | 1 - 2 | 65 | 99 |
NM_001288954.1 | NP_001275883.1 | 1 - 2 | 65 | 99 | |
GenBank mRNA | BF971643.1 | - | 1 - 2 | 44 | 99 |
BP317624.1 | - | 1 - 2 | 62 | 99 |