The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029858.3 | NP_001025029.2 | 5 - 6 | 996 | 79 |
XM_005266865.4 | XP_005266922.1 | 5 - 6 | 1296 | 79 | |
GenBank mRNA | AK128717.1 | - | 3 - 4 | 344 | 79 |
AK295860.1 | - | 5 - 6 | 693 | 79 | |
BC028615.1 | - | 5 - 6 | 999 | 79 | |
BC036493.1 | - | 5 - 6 | 764 | 79 | |
BC114925.1 | - | 6 - 7 | 870 | 79 | |
BX640761.1 | - | 2 - 3 | 257 | 79 |