Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
Chromosome Location
Chr.6: 75921114 - 76072678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282368.1 NP_001269297.1 15 - 16 2273 148
NM_001563.3 NP_001554.2 16 - 17 2507 148
GenBank mRNA AF047492.2 - 16 - 17 2447 148
BC117450.1 - 16 - 17 2337 148
BC117452.1 - 16 - 17 2337 148
CR749572.1 - 16 - 17 4749 148

View Full Product Details