Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC41
Entrez Gene ID
10489
Gene Name
leucine rich repeat containing 41
Gene Aliases
MUF1, PP7759
Chromosome Location
Chr.1: 46278400 - 46303366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006369.4 NP_006360.3 3 - 4 403 110
XM_017000074.1 XP_016855563.1 3 - 4 429 110
GenBank mRNA AF370430.1 - 3 - 4 302 111
AK024051.1 - 3 - 4 311 111
AK125827.1 - 2 - 3 1680 110
AK291283.1 - 3 - 4 386 110
AK296436.1 - 3 - 4 222 110
AK297596.1 - 3 - 4 417 110
BC092479.1 - 3 - 4 213 110
BC101522.1 - 3 - 4 393 110
BC101524.1 - 3 - 4 393 110
BM548142.1 - - 363 110
BX537366.1 - 3 - 4 313 111
CF125098.1 - 3 - 4 403 111

View Full Product Details