The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321307.1 | NP_001308236.1 | 1 - 2 | 136 | 78 |
NM_002519.2 | NP_002510.2 | 1 - 2 | 136 | 78 | |
XM_011542854.2 | XP_011541156.1 | 1 - 2 | 136 | 78 | |
GenBank mRNA | AK290020.1 | - | 1 - 2 | 136 | 78 |
AK291687.1 | - | 1 - 2 | 60 | 78 | |
BC040356.1 | - | 1 - 2 | 127 | 78 | |
BC050561.2 | - | 1 - 2 | 121 | 78 | |
BC136270.1 | - | 1 - 2 | 57 | 78 | |
D83243.1 | - | 1 - 2 | 100 | 78 | |
DA338219.1 | - | 1 - 2 | 136 | 78 | |
X97186.1 | - | 1 - 2 | 68 | 78 |