The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321307.1 | NP_001308236.1 | 3 - 4 | 323 | 150 |
NM_002519.2 | NP_002510.2 | 3 - 4 | 323 | 150 | |
XM_011542854.2 | XP_011541156.1 | 3 - 4 | 323 | 150 | |
XM_017017802.1 | XP_016873291.1 | 3 - 4 | 1061 | 150 | |
XM_017017803.1 | XP_016873292.1 | 3 - 4 | 504 | 150 | |
GenBank mRNA | AK290020.1 | - | 3 - 4 | 323 | 150 |
AK291687.1 | - | 3 - 4 | 247 | 150 | |
BC040356.1 | - | 3 - 4 | 314 | 150 | |
BC050561.2 | - | 3 - 4 | 308 | 150 | |
BC136270.1 | - | 3 - 4 | 244 | 150 | |
D83243.1 | - | 3 - 4 | 287 | 150 | |
DA338219.1 | - | 3 - 4 | 323 | 150 | |
U58852.1 | - | 3 - 4 | 221 | 150 | |
X97186.1 | - | 3 - 4 | 255 | 150 |