The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321307.1 | NP_001308236.1 | 5 - 6 | 433 | 73 |
NM_002519.2 | NP_002510.2 | 5 - 6 | 433 | 73 | |
XM_011542854.2 | XP_011541156.1 | 5 - 6 | 433 | 73 | |
XM_017017802.1 | XP_016873291.1 | 5 - 6 | 1171 | 73 | |
XM_017017803.1 | XP_016873292.1 | 5 - 6 | 614 | 73 | |
GenBank mRNA | AK290020.1 | - | 5 - 6 | 433 | 73 |
BC040356.1 | - | 5 - 6 | 424 | 73 | |
BC050561.2 | - | 5 - 6 | 418 | 73 | |
BC136270.1 | - | 5 - 6 | 354 | 73 | |
D83243.1 | - | 5 - 6 | 397 | 73 | |
DA338219.1 | - | 5 - 6 | 433 | 73 | |
U58852.1 | - | 5 - 6 | 331 | 73 | |
X97186.1 | - | 5 - 6 | 365 | 73 |