Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPAT
Entrez Gene ID
4863
Gene Name
nuclear protein, coactivator of histone transcription
Gene Aliases
E14, E14/NPAT, p220
Chromosome Location
Chr.11: 108155280 - 108222638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321307.1 NP_001308236.1 9 - 10 916 145
NM_002519.2 NP_002510.2 9 - 10 916 145
XM_011542854.2 XP_011541156.1 9 - 10 916 145
XM_017017802.1 XP_016873291.1 9 - 10 1654 145
XM_017017803.1 XP_016873292.1 9 - 10 1097 145
GenBank mRNA AK290020.1 - 9 - 10 916 145
AK291687.1 - 9 - 10 840 145
BC040356.1 - 9 - 10 907 145
BC050561.2 - 9 - 10 901 145
BC136270.1 - 9 - 10 837 145
D83243.1 - 9 - 10 880 145
U58852.1 - 9 - 10 814 145
X97186.1 - 9 - 10 848 145

View Full Product Details