The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152722.4 | NP_689935.2 | 3 - 4 | 1118 | 68 |
XM_005271449.2 | XP_005271506.1 | 3 - 4 | 1118 | 68 | |
XM_017017361.1 | XP_016872850.1 | 3 - 4 | 1118 | 68 | |
GenBank mRNA | AK098396.1 | - | 3 - 4 | 756 | 68 |
AK122595.1 | - | 3 - 4 | 756 | 68 | |
AY047587.1 | - | 3 - 4 | 756 | 68 | |
BC104831.1 | - | 3 - 4 | 740 | 68 | |
BC113562.1 | - | 3 - 4 | 740 | 68 |