The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147172.2 | NP_671701.1 | 1 - 2 | 88 | 61 |
GenBank mRNA | BG622439.1 | - | 1 - 2 | 42 | 61 |