Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZAR1
Entrez Gene ID
326340
Gene Name
zygote arrest 1
Gene Aliases
Z3CXXC6
Chromosome Location
Chr.4: 48490252 - 48497528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175619.2 NP_783318.1 2 - 3 1095 96
GenBank mRNA AY191416.1 - 2 - 3 1055 96
BC136265.1 - 2 - 3 1095 96

View Full Product Details