The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134363.2 | NP_001127835.2 | 2 - 3 | 1333 | 68 |
XM_011539697.2 | XP_011537999.1 | 2 - 3 | 1171 | 68 | |
XM_017016103.1 | XP_016871592.1 | 2 - 3 | 2591 | 68 | |
XM_017016104.1 | XP_016871593.1 | 2 - 3 | 4807 | 68 | |
GenBank mRNA | EU822950.1 | - | 2 - 3 | 1333 | 68 |