The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003647.2 | NP_003638.1 | 7 - 8 | 1282 | 135 |
XM_011525394.2 | XP_011523696.1 | 7 - 8 | 1260 | 135 | |
XM_011525395.2 | XP_011523697.1 | 7 - 8 | 1267 | 135 | |
XM_011525396.2 | XP_011523698.1 | 7 - 8 | 1269 | 135 | |
XM_017025243.1 | XP_016880732.1 | 7 - 8 | 1649 | 135 | |
XM_017025244.1 | XP_016880733.1 | 7 - 8 | 1263 | 135 | |
XM_017025245.1 | XP_016880734.1 | 7 - 8 | 1266 | 135 | |
GenBank mRNA | AB209365.1 | - | 6 - 7 | 932 | 135 |
AK293039.1 | - | 7 - 8 | 1229 | 135 | |
BC130629.1 | - | 6 - 7 | 1126 | 135 | |
BC130631.1 | - | 6 - 7 | 1126 | 135 | |
U49379.1 | - | 7 - 8 | 1189 | 135 |