The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024677.4 | NP_078953.3 | 11 - 12 | 2022 | 151 |
XM_017008611.1 | XP_016864100.1 | 10 - 11 | 1901 | 151 | |
XM_017008612.1 | XP_016864101.1 | 9 - 10 | 1799 | 151 | |
GenBank mRNA | AK094298.1 | - | 3 - 4 | 262 | 151 |
AK304048.1 | - | 11 - 12 | 1746 | 151 | |
BC042401.1 | - | 11 - 12 | 2022 | 151 |