The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205028.1 | NP_001191957.1 | 12 - 13 | 2008 | 91 |
NM_017440.4 | NP_059136.2 | 12 - 13 | 2113 | 91 | |
XM_005269026.1 | XP_005269083.1 | 13 - 14 | 2068 | 91 | |
XM_017019646.1 | XP_016875135.1 | 12 - 13 | 3414 | 91 | |
XM_017019647.1 | XP_016875136.1 | 14 - 15 | 2144 | 91 | |
XM_017019648.1 | XP_016875137.1 | 13 - 14 | 2575 | 91 | |
XM_017019649.1 | XP_016875138.1 | 13 - 14 | 2005 | 91 | |
XM_017019650.1 | XP_016875139.1 | 12 - 13 | 3925 | 91 | |
GenBank mRNA | AF007130.1 | - | 4 - 5 | 595 | 91 |
AK026745.1 | - | 7 - 8 | 1170 | 91 | |
AK297311.1 | - | 12 - 13 | 1913 | 91 | |
BC028355.2 | - | 12 - 13 | 2064 | 91 | |
BC034945.1 | - | 11 - 12 | 1658 | 91 |