Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MEG3
- Entrez Gene ID
-
55384
- Gene Name
- maternally expressed 3 (non-protein coding)
- Gene Aliases
-
FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
- Chromosome Location
-
Chr.14: 100826108 - 100861026 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033359.1 |
|
4 - 5 |
1012 |
87 |
View Full Product Details