The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015306.2 | NP_056121.2 | 5 - 6 | 1080 | 92 |
XM_017000831.1 | XP_016856320.1 | 5 - 6 | 1085 | 92 | |
XM_017000832.1 | XP_016856321.1 | 5 - 6 | 1082 | 92 | |
XM_017000833.1 | XP_016856322.1 | 5 - 6 | 577 | 92 | |
XM_017000834.1 | XP_016856323.1 | 5 - 6 | 577 | 92 | |
XM_017000835.1 | XP_016856324.1 | 5 - 6 | 1088 | 92 | |
XM_017000836.1 | XP_016856325.1 | 5 - 6 | 1088 | 92 | |
XM_017000837.1 | XP_016856326.1 | 5 - 6 | 1088 | 92 |