The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015718.2 | NP_056533.1 | 11 - 12 | 1562 | 118 |
GenBank mRNA | AF190122.1 | - | 11 - 12 | 1562 | 118 |