The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015718.2 | NP_056533.1 | 2 - 3 | 252 | 88 |
GenBank mRNA | AF190122.1 | - | 2 - 3 | 252 | 88 |