Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB11
Entrez Gene ID
27107
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
ZNF-U69274, ZNF913
Chromosome Location
Chr.3: 101649439 - 101677144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014415.3 NP_055230.2 8 - 9 2533 97
XM_011512689.2 XP_011510991.1 8 - 9 2241 97
XM_011512690.2 XP_011510992.1 6 - 7 3918 97
GenBank mRNA AB208862.1 - 5 - 6 1246 97
AK312859.1 - 8 - 9 2525 97
U69274.1 - 8 - 9 2464 97

View Full Product Details