The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324338.1 | NP_001311267.1 | 7 - 8 | 903 | 155 |
NM_152597.4 | NP_689810.3 | 7 - 8 | 1000 | 155 | |
XM_011521305.2 | XP_011519607.1 | 7 - 8 | 876 | 155 | |
XM_011521306.1 | XP_011519608.1 | 7 - 8 | 1289 | 155 | |
XM_011521307.2 | XP_011519609.1 | 7 - 8 | 876 | 155 | |
XM_011521309.2 | XP_011519611.1 | 7 - 8 | 876 | 155 | |
XM_011521311.2 | XP_011519613.1 | 3 - 4 | 1560 | 155 | |
XM_017021972.1 | XP_016877461.1 | 3 - 4 | 353 | 155 | |
GenBank mRNA | AK093308.1 | - | 7 - 8 | 982 | 155 |
AY260140.1 | - | 6 - 7 | 797 | 155 | |
BC045191.2 | - | 7 - 8 | 992 | 155 |