Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANBA
Entrez Gene ID
4126
Gene Name
mannosidase beta
Gene Aliases
MANB1
Chromosome Location
Chr.4: 102631486 - 102760998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005908.3 NP_005899.3 2 - 3 375 143
XM_017008203.1 XP_016863692.1 3 - 4 361 143
XM_017008204.1 XP_016863693.1 2 - 3 330 143
GenBank mRNA AB209846.1 - 2 - 3 322 143
AK291598.1 - 2 - 3 363 143
AK300015.1 - 2 - 3 316 143
AK312827.1 - 2 - 3 330 143
AU132611.1 - 2 - 3 375 143
BC015743.1 - 2 - 3 365 143
EU009130.1 - 2 - 3 275 143
U60337.1 - 2 - 3 362 143

View Full Product Details