Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEGFC
Entrez Gene ID
7424
Gene Name
vascular endothelial growth factor C
Gene Aliases
Flt4-L, LMPH1D, VRP
Chromosome Location
Chr.4: 176683534 - 176792745 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005429.4 NP_005420.1 6 - 7 1578 93
GenBank mRNA AK313879.1 - 6 - 7 1344 93
BC035212.1 - 6 - 7 1409 93
BC063685.1 - 6 - 7 1560 93
CR541897.1 - 6 - 7 1144 93
U43142.1 - 6 - 7 1515 93
U58111.1 - 6 - 7 1467 93
X94216.1 - 6 - 7 1495 93

View Full Product Details