The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321729.1 | NP_001308658.1 | 12 - 13 | 1377 | 153 |
NM_001321730.1 | NP_001308659.1 | 12 - 13 | 1377 | 153 | |
NM_001321731.1 | NP_001308660.1 | 12 - 13 | 1377 | 153 | |
NM_001321733.1 | NP_001308662.1 | 12 - 13 | 1377 | 153 | |
NM_001321734.1 | NP_001308663.1 | 11 - 12 | 1211 | 153 | |
NM_015189.2 | NP_056004.1 | 12 - 13 | 1377 | 153 | |
NR_135773.1 | 12 - 13 | 1377 | 153 | ||
NR_135774.1 | 12 - 13 | 1377 | 153 | ||
XM_005264224.1 | XP_005264281.1 | 5 - 6 | 669 | 153 | |
XM_011532711.2 | XP_011531013.1 | 12 - 13 | 1373 | 153 | |
XM_011532712.2 | XP_011531014.1 | 12 - 13 | 1373 | 153 | |
XM_017003641.1 | XP_016859130.1 | 8 - 9 | 811 | 153 | |
XM_017003642.1 | XP_016859131.1 | 6 - 7 | 424 | 153 | |
GenBank mRNA | AB023136.1 | - | 12 - 13 | 1360 | 153 |
AK026474.1 | - | 9 - 10 | 872 | 153 |