Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSTK
Entrez Gene ID
118672
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
C10orf89
Chromosome Location
Chr.10: 122980040 - 122997519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153336.2 NP_699167.2 4 - 5 1226 140
XM_017015640.1 XP_016871129.1 4 - 5 852 140
GenBank mRNA AK094271.1 - 3 - 4 1331 140
BC035344.1 - 4 - 5 1226 140

View Full Product Details