Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO7B
Entrez Gene ID
4648
Gene Name
myosin VIIB
Gene Aliases
-
Chromosome Location
Chr.2: 127535689 - 127637728 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080527.1 NP_001073996.1 3 - 4 183 78
XM_006712539.3 XP_006712602.1 3 - 4 296 78
XM_011511218.2 XP_011509520.1 3 - 4 296 78
XM_017004169.1 XP_016859658.1 3 - 4 296 78

View Full Product Details